Assay Design
Gene Transcripts
- Gene Symbol
- Sdr9c7
- Entrez Gene ID
-
70061
- Gene Name
- 4short chain dehydrogenase/reductase family 9C, member 7
- Gene Aliases
-
1810054F20Rik, Rdh20, Rdhs, SDR-O, Sdro
- Chromosome Location
-
Chr.10: 127898535 - 127911761 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY044433.1 |
- |
2 - 3 |
642 |
79 |
View Full Product Details