Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028476.4 | NP_082752.3 | 7 - 8 | 612 | 100 |
GenBank mRNA | AK034725.1 | - | 7 - 8 | 590 | 100 |
AK042804.1 | - | 2 - 3 | 107 | 100 | |
AK049682.1 | - | 7 - 8 | 562 | 100 | |
BC030669.1 | - | 7 - 8 | 590 | 100 |