Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028994.2 | NP_083270.1 | 1 - 2 | 253 | 64 |
GenBank mRNA | AK030501.1 | - | 1 - 2 | 254 | 64 |
AK031612.1 | - | 1 - 2 | 98 | 64 | |
AK034927.1 | - | 1 - 2 | 132 | 64 | |
AK147972.1 | - | 1 - 2 | 107 | 64 | |
AK161944.1 | - | 1 - 2 | 133 | 64 | |
AK165094.1 | - | 1 - 2 | 131 | 64 | |
AK170242.1 | - | 1 - 2 | 107 | 64 | |
BC010318.1 | - | 1 - 2 | 116 | 64 | |
BC011126.1 | - | 1 - 2 | 113 | 64 |