Product Details

Assay Reference Genome
Location

Chr.4:63429300 on build GRCm38
Cytoband
4C1
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
73750
Gene Symbol
Whrn
Gene Name
whirlin
Gene Aliases
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi
Location
Chr.4:63414909-63496078 on build GRCm38
Assay Gene Location
Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Whrn NM_001008791.2 NP_001008791.1
NM_001008792.2 NP_001008792.1
NM_001008793.2 NP_001008793.1
NM_028640.2 NP_082916.1
AK021254.1 BAB32350.1
AK122523.1 BAC65805.1
AK190484.1 2 198
AY739114.1 AAV87519.1
AY739115.1 AAV87520.1
AY739116.1 AAV87521.1
AY739117.1 AAV87522.1
AY739118.1 AAV87523.1
AY739119.1 AAV87524.1
AY739120.1 AAV87525.1
AY739121.1 AAV87526.1
AY739122.1 3 986 AAV87527.1
BC150757.1
HQ148552.1

View Full Product Details