Assay Design



Gene Transcripts

Gene Symbol
Itih5
Entrez Gene ID
209378
Gene Name
inter-alpha (globulin) inhibitor H5
Gene Aliases
4631408O11Rik, 5430408M01Rik, E130106B02
Chromosome Location
Chr.2: 10153536 - 10256529 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172471.2 NP_766059.1 5 - 6 776 114
GenBank mRNA AK077381.1 - - 699 114
AK087428.1 - 5 - 6 704 114
AK134707.1 - 5 - 6 704 114
AK140756.1 - 5 - 6 777 114
BC043314.1 - 5 - 6 719 114
BC062196.1 - 5 - 6 760 114

View Full Product Details