Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172488.2 | NP_766076.1 | 6 - 7 | 1483 | 81 |
GenBank mRNA | AK033565.1 | - | 6 - 7 | 1484 | 81 |
AK152154.1 | - | 6 - 7 | 1424 | 81 | |
AK153349.1 | - | 6 - 7 | 1508 | 81 | |
BC116748.1 | - | 5 - 6 | 1239 | 81 | |
BC119140.1 | - | 5 - 6 | 1239 | 81 |