Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172507.5 | NP_766095.1 | 1 - 2 | 162 | 59 |
GenBank mRNA | AK032092.1 | - | - | 175 | 59 |
AK076202.1 | - | 1 - 2 | 151 | 59 | |
AK160357.1 | - | 1 - 2 | 151 | 59 | |
BC038473.1 | - | 1 - 2 | 201 | 59 |