Assay Design



Gene Transcripts

Gene Symbol
Nutm1
Entrez Gene ID
213765
Gene Name
NUT midline carcinoma, family member 1
Gene Aliases
4932438M10, Nut
Chromosome Location
Chr.2: 112247948 - 112259291 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172521.1 NP_766109.1 2 - 3 836 88
GenBank mRNA AK077049.1 - 2 - 3 836 88
BC125332.1 - 2 - 3 816 88
BC125334.1 - 2 - 3 816 88

View Full Product Details