Assay Design



Gene Transcripts

Gene Symbol
Megf11
Entrez Gene ID
214058
Gene Name
multiple EGF-like-domains 11
Gene Aliases
2410080H04Rik, D130061K05
Chromosome Location
Chr.9: 64385618 - 64709205 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134399.1 NP_001127871.1 3 - 4 407 62
NM_172522.4 NP_766110.3 3 - 4 407 62
GenBank mRNA AK032661.1 - 2 - 3 126 62
AK048840.1 - 3 - 4 351 62
AK051642.1 - 3 - 4 344 62
AK122555.2 - 3 - 4 347 62
BB650216.1 - 3 - 4 348 62
BC151157.1 - 3 - 4 308 62
BC172024.1 - 3 - 4 308 62
CJ049881.1 - 3 - 4 383 62

View Full Product Details