Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172523.3 | NP_766111.1 | 13 - 14 | 1351 | 81 |
GenBank mRNA | AJ555564.1 | - | 12 - 13 | 1215 | 81 |
AK033871.1 | - | 13 - 14 | 1307 | 81 | |
AK034368.1 | - | 8 - 9 | 2710 | 81 | |
AK035644.1 | - | 13 - 14 | 1335 | 81 | |
AK041274.1 | - | 13 - 14 | 1303 | 81 | |
BC078449.1 | - | 12 - 13 | 1222 | 81 |