Assay Design



Gene Transcripts

Gene Symbol
Nipal4
Entrez Gene ID
214112
Gene Name
NIPA-like domain containing 4
Gene Aliases
9530066K23Rik
Chromosome Location
Chr.11: 46148155 - 46166359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172524.3 NP_766112.1 1 - 2 125 78
GenBank mRNA AK035561.1 - 1 - 2 126 78
AK035736.1 - 1 - 2 102 78
BC139819.1 - 1 - 2 116 78

View Full Product Details