Assay Design



Gene Transcripts

Gene Symbol
Ehmt1
Entrez Gene ID
77683
Gene Name
euchromatic histone methyltransferase 1
Gene Aliases
9230102N17Rik, D330003E03, Eu-HMTase1, GLP, GLP1, KMT1D, mKIAA1876
Chromosome Location
Chr.2: 24790769 - 24919643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012518.3 NP_001012536.2 2 - 3 123 69
NM_001109686.2 NP_001103156.1 2 - 3 123 69
NM_001109687.2 NP_001103157.1 2 - 3 123 69
NM_172545.4 NP_766133.2 2 - 3 123 69
GenBank mRNA AB205007.1 - 2 - 3 129 69
AK052174.1 - 2 - 3 105 69
AK054555.1 - 2 - 3 91 69
AK082062.1 - 2 - 3 113 69
AK084994.1 - 2 - 3 106 69
BB621957.1 - 2 - 3 121 69

View Full Product Details