Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172564.3 | NP_766152.2 | 7 - 8 | 1700 | 78 |
GenBank mRNA | AK036834.1 | - | 7 - 8 | 1697 | 78 |
AK039855.1 | - | 2 - 3 | 326 | 78 | |
AK170638.1 | - | 1 - 2 | 72 | 78 | |
BC055820.1 | - | 7 - 8 | 1682 | 78 |