Assay Design



Gene Transcripts

Gene Symbol
Pggt1b
Entrez Gene ID
225467
Gene Name
protein geranylgeranyltransferase type I, beta subunit
Gene Aliases
2010207C17Rik, 2610100E13, AI451237, AI551093, BGG1, GGT1
Chromosome Location
Chr.18: 46235474 - 46281089 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172627.3 NP_766215.1 4 - 5 512 61
GenBank mRNA AK028188.1 - 4 - 5 371 61
AK081731.1 - 4 - 5 513 61
BC141021.1 - 4 - 5 510 61

View Full Product Details