Assay Design



Gene Transcripts

Gene Symbol
Sh3rf3
Entrez Gene ID
237353
Gene Name
SH3 domain containing ring finger 3
Gene Aliases
4831416G18Rik, Igfbp5-ip, Sh3md4, Sh3rf31
Chromosome Location
Chr.10: 58812372 - 59138916 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172788.3 NP_766376.2 4 - 5 1502 107
GenBank mRNA AK029202.1 - 4 - 5 1405 107
BC057304.1 - 4 - 5 758 107

View Full Product Details