Assay Design



Gene Transcripts

Gene Symbol
Palm2
Entrez Gene ID
242481
Gene Name
paralemmin 2
Gene Aliases
53-H12, B130066I24, R75368, pEN70
Chromosome Location
Chr.4: 57568248 - 57717146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172868.3 NP_766456.1 2 - 3 247 67
GenBank mRNA AK045344.1 - 2 - 3 248 67
AK133666.1 - - 208 67
BC113156.1 - 2 - 3 213 67

View Full Product Details