Assay Design
Gene Transcripts
- Gene Symbol
- Pramef8
- Entrez Gene ID
-
242736
- Gene Name
- PRAME family member 8
- Gene Aliases
-
4732496O08Rik, AU023234
- Chromosome Location
-
Chr.4: 143412309 - 143421091 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_172877.2 |
NP_766465.1 |
2 - 3 |
729 |
125 |
GenBank mRNA |
AK029142.1 |
- |
2 - 3 |
730 |
125 |
View Full Product Details