Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172948.3 | NP_766536.2 | 15 - 16 | 2461 | 68 |
GenBank mRNA | AB116028.1 | - | 15 - 16 | 1848 | 68 |
AB119127.1 | - | 15 - 16 | 1927 | 68 | |
AK049266.1 | - | 4 - 5 | 780 | 68 | |
BC063084.1 | - | 14 - 15 | 2362 | 68 | |
BC094604.1 | - | 15 - 16 | 2461 | 68 |