Assay Design
Gene Transcripts
- Gene Symbol
- Sh3rf2
- Entrez Gene ID
-
269016
- Gene Name
- SH3 domain containing ring finger 2
- Gene Aliases
-
2310046K19, 9130023G24Rik, AI452083, Ppp1r39, RNF158
- Chromosome Location
-
Chr.18: 42051743 - 42158695 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_172966.3 |
NP_766554.2 |
2 - 3 |
757 |
106 |
GenBank mRNA |
AK029168.1 |
- |
2 - 3 |
758 |
106 |
View Full Product Details