Assay Design
Gene Transcripts
- Gene Symbol
- Ip6k3
- Entrez Gene ID
-
271424
- Gene Name
- inositol hexaphosphate kinase 3
- Gene Aliases
-
D830007E07Rik, Ihpk3
- Chromosome Location
-
Chr.17: 27143971 - 27167764 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173027.2 |
NP_766615.1 |
1 - 2 |
122 |
115 |
GenBank mRNA |
AK052857.1 |
- |
1 - 2 |
123 |
115 |
View Full Product Details