Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173410.3 | NP_775586.1 | 2 - 3 | 1252 | 65 |
GenBank mRNA | AK036100.1 | - | 2 - 3 | 1255 | 65 |
AK042755.1 | - | 2 - 3 | 354 | 65 | |
AY255585.1 | - | 2 - 3 | 436 | 65 | |
BC119380.1 | - | 2 - 3 | 1110 | 65 | |
BC120647.1 | - | 2 - 3 | 1110 | 65 |