Assay Design



Gene Transcripts

Gene Symbol
Fkrp
Entrez Gene ID
243853
Gene Name
fukutin related protein
Gene Aliases
A830029B19Rik, AI842067, AI847300, LGMD1I, MDC1C
Chromosome Location
Chr.7: 16809267 - 16820668 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173430.2 NP_775606.1 2 - 3 96 68
GenBank mRNA AJ511806.1 - 2 - 3 79 68
AK135089.1 - 2 - 3 97 68
AK166068.1 - 2 - 3 98 68
AK168308.1 - 2 - 3 94 68
BC029216.1 - 2 - 3 91 68
BC053072.1 - 2 - 3 79 68

View Full Product Details