Assay Design



Gene Transcripts

Gene Symbol
Impg2
Entrez Gene ID
224224
Gene Name
interphotoreceptor matrix proteoglycan 2
Gene Aliases
IPM200, PG10.2, Rsbp, Spacrcan
Chromosome Location
Chr.16: 56203842 - 56273753 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174876.3 NP_777365.2 4 - 5 731 105
GenBank mRNA AY174061.1 - 4 - 5 731 105
BC048863.1 - 4 - 5 700 105

View Full Product Details