Assay Design
Gene Transcripts
- Gene Symbol
- Agtr1b
- Entrez Gene ID
-
11608
- Gene Name
- angiotensin II receptor, type 1b
- Gene Aliases
-
AT1B, AT2R1B, Agtr-1b, Angtr-1b
- Chromosome Location
-
Chr.3: 20314473 - 20367177 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175086.3 |
NP_780295.2 |
2 - 2 |
532 |
66 |
GenBank mRNA |
AK087228.1 |
- |
2 - 2 |
532 |
66 |
- Gene Symbol
- Agtr1a
- Entrez Gene ID
-
11607
- Gene Name
- angiotensin II receptor, type 1a
- Gene Aliases
-
1810074K20Rik, AG2S, AI551199, AT1, AT1a, AT2R1, AT2R1A, Agtr-1a, Agtr1, Angtr-1a
- Chromosome Location
-
Chr.13: 30336356 - 30382867 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177322.3 |
NP_796296.1 |
3 - 3 |
|
66 |
GenBank mRNA |
BC036175.1 |
- |
3 - 3 |
|
66 |
View Full Product Details