Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252330.1 | NP_001239259.1 | 8 - 9 | 1680 | 84 |
NM_175328.3 | NP_780537.1 | 9 - 10 | 1777 | 84 | |
GenBank mRNA | AK036136.1 | - | 8 - 9 | 1659 | 84 |
AM085111.1 | - | 7 - 8 | 1308 | 84 | |
AY149280.1 | - | 9 - 10 | 1742 | 84 | |
AY149281.1 | - | 8 - 9 | 1655 | 84 | |
BC076593.1 | - | 8 - 9 | 1680 | 84 |