Assay Design



Gene Transcripts

Gene Symbol
Coro2b
Entrez Gene ID
235431
Gene Name
coronin, actin binding protein, 2B
Gene Aliases
CLIPINC, E130012P22Rik
Chromosome Location
Chr.9: 62419490 - 62537044 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175484.3 NP_780693.2 3 - 4 714 65
GenBank mRNA AK047527.1 - 3 - 4 656 65
AK087414.1 - 3 - 4 715 65
BC062649.1 - 3 - 4 446 65

View Full Product Details