Assay Design
Gene Transcripts
- Gene Symbol
- Plod1
- Entrez Gene ID
-
18822
- Gene Name
- procollagen-lysine, 2-oxoglutarate 5-dioxygenase 1
- Gene Aliases
-
2410042F05Rik, AI854890, AV007929, Lh1, Plod
- Chromosome Location
-
Chr.4: 147909753 - 147936776 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF046782.1 |
- |
17 - 18 |
1937 |
58 |
BC010268.1 |
- |
9 - 10 |
954 |
58 |
View Full Product Details