Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013739.2 | NP_038767.1 | 1 - 2 | 90 | 59 |
GenBank mRNA | AF179242.1 | - | 1 - 2 | 90 | 59 |
AF237580.1 | - | 1 - 2 | 86 | 59 | |
AK137520.1 | - | 1 - 2 | 77 | 59 | |
BC116922.1 | - | 1 - 2 | 89 | 59 | |
BC120535.1 | - | 1 - 2 | 89 | 59 |