Assay Design



Gene Transcripts

Gene Symbol
Jak1
Entrez Gene ID
16451
Gene Name
Janus kinase 1
Gene Aliases
AA960307, BAP004, C130039L05Rik
Chromosome Location
Chr.4: 101151974 - 101265297 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146145.2 NP_666257.2 14 - 15 2306 80
GenBank mRNA AK133866.1 - 14 - 15 2249 80
AK141210.1 - 14 - 15 2248 80
AK143650.1 - 14 - 15 2253 80
AK148521.1 - 14 - 15 2270 80
AK152128.1 - 14 - 15 2269 80
BC024100.1 - 6 - 7 716 80
BC031297.1 - 4 - 5 427 80
S63728.1 - 14 - 15 2256 80

View Full Product Details