Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114311.1 | NP_001107783.1 | 2 - 3 | 1676 | 86 |
NM_001286376.1 | NP_001273305.1 | 2 - 3 | 1676 | 86 | |
NM_175162.4 | NP_780371.3 | 2 - 3 | 471 | 86 | |
GenBank mRNA | AK030015.1 | - | 2 - 3 | 285 | 86 |
AK053894.1 | - | 2 - 3 | 1676 | 86 | |
AK129346.1 | - | 2 - 3 | 1636 | 86 | |
AK162847.1 | - | 2 - 3 | 505 | 86 | |
BC100572.1 | - | 2 - 3 | 471 | 86 | |
BC100573.1 | - | 2 - 3 | 471 | 86 | |
BC138237.1 | - | 2 - 3 | 242 | 86 | |
BC138238.1 | - | 2 - 3 | 242 | 86 |