Assay Design



Gene Transcripts

Gene Symbol
Wdr27
Entrez Gene ID
71682
Gene Name
WD repeat domain 27
Gene Aliases
0610012K18Rik
Chromosome Location
Chr.17: 14784832 - 14943151 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175173.3 NP_780382.2 20 - 21 2039 90
GenBank mRNA AK040117.1 - 5 - 6 479 90
AK077050.1 - 20 - 21 2040 90

View Full Product Details