Assay Design



Gene Transcripts

Gene Symbol
Wdr27
Entrez Gene ID
71682
Gene Name
WD repeat domain 27
Gene Aliases
0610012K18Rik
Chromosome Location
Chr.17: 14784832 - 14943151 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175173.3 NP_780382.2 23 - 24 2360 87
GenBank mRNA AK040117.1 - 8 - 9 800 87
AK077050.1 - 23 - 24 2369 87
AV277048.2 - 3 - 4 278 87

View Full Product Details