Assay Design



Gene Transcripts

Gene Symbol
Klhl5
Entrez Gene ID
71778
Gene Name
kelch-like 5
Gene Aliases
1300013C10Rik
Chromosome Location
Chr.5: 65107527 - 65168186 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175174.4 NP_780383.2 2 - 3 731 60
GenBank mRNA AK004982.1 - 2 - 3 467 60
AK090246.1 - 2 - 3 291 60
AK220195.1 - 2 - 3 477 60
BC057606.1 - 2 - 3 731 60

View Full Product Details