Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175360.2 | NP_780569.1 | 1 - 2 | 213 | 64 |
GenBank mRNA | AK075574.1 | - | 1 - 2 | 213 | 64 |
AK135228.1 | - | 1 - 2 | 231 | 64 | |
AK146929.1 | - | 1 - 2 | 291 | 64 | |
AK164386.1 | - | 1 - 2 | 223 | 64 | |
AK166348.1 | - | 1 - 2 | 287 | 64 | |
BC029548.1 | - | 1 - 2 | 253 | 64 |