Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175370.4 | NP_780579.1 | 11 - 12 | 999 | 103 |
GenBank mRNA | AK077190.1 | - | 11 - 12 | 967 | 103 |
AK133373.1 | - | - | 997 | 103 | |
BC145039.1 | - | 10 - 11 | 906 | 103 | |
BC145040.1 | - | 10 - 11 | 904 | 103 |