Assay Design



Gene Transcripts

Gene Symbol
Fbxw17
Entrez Gene ID
109082
Gene Name
F-box and WD-40 domain protein 17
Gene Aliases
1110064L07Rik, C85886
Chromosome Location
Chr.13: 50417834 - 50433781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175401.3 NP_780610.2 9 - 10 1310 83
GenBank mRNA AK075676.1 - 1 - 2 1435 83
AK155157.1 - 10 - 11 1824 83
BC040428.1 - 9 - 10 1310 83

View Full Product Details