Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039493.1 | NP_001034582.1 | 5 - 6 | 2249 | 77 |
GenBank mRNA | AK034959.1 | - | 5 - 6 | 2171 | 77 |
AK172220.1 | - | 4 - 5 | 1342 | 77 | |
BC063098.1 | - | - | 2249 | 77 |