Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177387.3 | NP_796361.2 | 4 - 5 | 952 | 115 |
GenBank mRNA | AK086181.1 | - | 4 - 5 | 899 | 115 |
AK147850.1 | - | 4 - 5 | 953 | 115 | |
AK162950.1 | - | 4 - 5 | 525 | 115 | |
BC138155.1 | - | 4 - 5 | 692 | 115 |