Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178374.3 | NP_848461.1 | 2 - 3 | 191 | 61 |
GenBank mRNA | AK013151.1 | - | 2 - 3 | 201 | 61 |
AK155315.1 | - | 2 - 3 | 208 | 61 | |
BC023317.1 | - | - | 171 | 61 | |
BC049638.1 | - | 2 - 3 | 191 | 61 | |
BC061258.1 | - | 2 - 3 | 196 | 61 |