Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178593.3 | NP_848708.2 | 2 - 3 | 310 | 75 |
GenBank mRNA | AK133961.1 | - | 2 - 3 | 265 | 75 |
AK158429.1 | - | 2 - 3 | 251 | 75 | |
AK171914.1 | - | 2 - 3 | 233 | 75 | |
BC025872.1 | - | 2 - 3 | 230 | 75 |