Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178654.4 | NP_848769.2 | 2 - 3 | 690 | 107 |
GenBank mRNA | AK028469.1 | - | 2 - 3 | 677 | 107 |
AK046248.1 | - | 2 - 3 | 607 | 107 | |
AK049713.1 | - | 2 - 3 | 677 | 107 | |
AK077910.1 | - | 2 - 3 | 670 | 107 | |
AK133298.1 | - | 2 - 3 | 679 | 107 | |
AK171092.1 | - | 2 - 3 | 621 | 107 | |
BC052073.1 | - | 3 - 4 | 613 | 107 |