Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184706.1 | NP_001171635.1 | 1 - 2 | 284 | 88 |
NM_001184708.1 | NP_001171637.1 | 1 - 2 | 284 | 88 | |
NM_178667.4 | NP_848782.2 | 1 - 2 | 284 | 88 | |
GenBank mRNA | AK029055.1 | - | 1 - 2 | 257 | 88 |
AK044784.1 | - | 1 - 2 | 286 | 88 | |
AK053881.1 | - | 1 - 2 | 242 | 88 | |
AK079624.1 | - | 1 - 2 | 212 | 88 | |
AK162972.1 | - | 1 - 2 | 242 | 88 |