Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178681.4 | NP_848796.2 | 11 - 12 | 1449 | 68 |
GenBank mRNA | AK032509.1 | - | 11 - 12 | 1608 | 68 |
AK038544.1 | - | 11 - 12 | 1177 | 68 | |
AK044516.1 | - | 11 - 12 | 1259 | 68 | |
AK047682.1 | - | 12 - 13 | 1385 | 68 | |
AK083222.1 | - | 12 - 13 | 1460 | 68 | |
AK122355.1 | - | 10 - 11 | 937 | 68 | |
AK139408.1 | - | 11 - 12 | 1450 | 68 | |
BC070461.1 | - | 11 - 12 | 1281 | 68 |