Assay Design



Gene Transcripts

Gene Symbol
Dpy19l3
Entrez Gene ID
233115
Gene Name
dpy-19-like 3 (C. elegans)
Gene Aliases
6030410G08, 9330164H19Rik
Chromosome Location
Chr.7: 35685500 - 35754454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178704.3 NP_848819.2 15 - 16 1859 64
GenBank mRNA AF538322.1 - 15 - 16 1843 64
AK033395.1 - 15 - 16 1816 64
AK045758.1 - 15 - 16 1760 64
BC050065.1 - 15 - 16 1803 64
BC059897.1 - 15 - 16 1755 64

View Full Product Details