Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178704.3 | NP_848819.2 | 3 - 4 | 477 | 78 |
GenBank mRNA | AF538322.1 | - | 3 - 4 | 461 | 78 |
AK031345.1 | - | 3 - 4 | 469 | 78 | |
AK033395.1 | - | 3 - 4 | 434 | 78 | |
AK034217.1 | - | 3 - 4 | 478 | 78 | |
AK045758.1 | - | 3 - 4 | 378 | 78 | |
BC050065.1 | - | 3 - 4 | 421 | 78 | |
BC059897.1 | - | 3 - 4 | 374 | 78 |