Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271915.1 | NP_001258844.1 | 1 - 1 | 1852 | 64 |
NM_178879.4 | NP_849210.1 | 2 - 2 | 1664 | 64 | |
GenBank mRNA | AK032108.1 | - | 1 - 1 | 1853 | 64 |
AK053867.1 | - | 1 - 1 | 1366 | 64 | |
AK169360.1 | - | 1 - 1 | 1848 | 64 | |
BC047209.1 | - | 1 - 1 | 1824 | 64 | |
BC108347.1 | - | 1 - 1 | 1802 | 64 | |
BC120583.1 | - | 1 - 1 | 1517 | 64 | |
BC120585.1 | - | 1 - 1 | 1517 | 64 |