Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181328.3 | NP_851845.1 | 2 - 3 | 59 | |
GenBank mRNA | AK047628.1 | - | 2 - 3 | 59 | |
AK087057.1 | - | 2 - 3 | 59 | ||
AK148350.1 | - | 2 - 3 | 251 | 59 | |
AK160906.1 | - | 2 - 3 | 59 | ||
BC006711.1 | - | 2 - 3 | 146 | 59 |