Assay Design



Gene Transcripts

Gene Symbol
Pik3c3
Entrez Gene ID
225326
Gene Name
phosphoinositide-3-kinase, class 3
Gene Aliases
5330434F23Rik, Vps34
Chromosome Location
Chr.18: 30272094 - 30348126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181414.5 NP_852079.2 22 - 23 2489 77
GenBank mRNA AK044997.1 - 22 - 23 2488 77
BC024675.1 - 22 - 23 2521 77
BC027029.1 - 22 - 23 2531 77
BC057678.1 - 22 - 23 2492 77

View Full Product Details