Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133972.2 | NP_598733.2 | 3 - 4 | 526 | 72 |
GenBank mRNA | AK010815.1 | - | 3 - 4 | 554 | 72 |
AK049351.1 | - | 3 - 4 | 242 | 72 | |
AK080609.1 | - | 3 - 4 | 231 | 72 | |
AK081550.1 | - | 3 - 4 | 571 | 72 | |
BC043070.1 | - | 3 - 4 | 526 | 72 | |
BC048396.1 | - | 3 - 4 | 487 | 72 | |
BC055779.1 | - | 3 - 4 | 526 | 72 |