Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173775.3 | NP_776136.2 | 16 - 17 | 1888 | 62 |
GenBank mRNA | AK048790.1 | - | 8 - 9 | 2483 | 62 |
BC047211.1 | - | 14 - 15 | 1570 | 62 | |
BC079632.1 | - | 4 - 5 | 616 | 62 |